Merck
  • MATILDE chemotherapy regimen for primary CNS lymphoma: results at a median follow-up of 12 years.
MATILDE chemotherapy regimen for primary CNS lymphoma: results at a median follow-up of 12 years.
Neurology (2014-03-19)
Andrés J M Ferreri, Fabio Ciceri, Alba A Brandes, Mauro Montanari, Monica Balzarotti, Michele Spina, Fiorella Ilariucci, Francesco Zaja, Caterina Stelitano, Flavio Bobbio, Gaetano Corazzelli, Luca Baldini, Michele Reni
ABSTRACT
We report updated results at a median follow-up of 12 years of a phase II trial assessing first-line MATILDE chemotherapy and response-tailored radiotherapy in patients with primary CNS lymphomas (PCNSL). Forty-one HIV-negative patients (18-70 years; Eastern Cooperative Oncology Group performance status ≤3) with histologically confirmed PCNSL received 3 courses of MATILDE chemotherapy followed by whole-brain radiotherapy (WBRT). Chemotherapy activity was the primary endpoint. Overall response rate was 76% (95% confidence interval [CI] 63%-89%) after chemotherapy and 83% (95% CI 71%-95%) after chemotherapy ± radiotherapy. At a median follow-up of 144 months (range 47-153), 31 patients experienced an event: relapse in 24, progressive disease in 3, and toxic death in 4, with a 5-year progression-free survival of 24% ± 8%. Two patients experienced a late relapse (100 and 101 months). Nine patients are alive and disease-free, 8 of whom are alive at >10 years, with a 5-year overall survival of 30% ± 7%. At 10 years from diagnosis, no patient showed chronic hematologic and nonhematologic toxicities, with a Mini-Mental State Examination score of ≥29 in all cases but one. At a median follow-up of 12 years, MATILDE regimen followed by WBRT confirmed the previously reported survival plateau, which further proves its long-lasting efficacy with acceptable neurologic deficits. This study provides Class IV evidence that in patients with PCNSL, MATILDE chemotherapy followed by response-tailored radiotherapy increases the probability of disease remission at 12 years.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thio-TEPA
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Idarubicin hydrochloride, solid
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
SAFC
Methotrexate
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material