Merck
  • Human UGT1A4 and UGT1A3 conjugate 25-hydroxyvitamin D3: metabolite structure, kinetics, inducibility, and interindividual variability.
Human UGT1A4 and UGT1A3 conjugate 25-hydroxyvitamin D3: metabolite structure, kinetics, inducibility, and interindividual variability.
Endocrinology (2014-03-20)
Zhican Wang, Timothy Wong, Takanori Hashizume, Leslie Z Dickmann, Michele Scian, Nicholas J Koszewski, Jesse P Goff, Ronald L Horst, Amarjit S Chaudhry, Erin G Schuetz, Kenneth E Thummel
ABSTRACT
25-Hydroxyvitamin D3 (25OHD3) is used as a clinical biomarker for assessment of vitamin D status. Blood levels of 25OHD3 represent a balance between its formation rate and clearance by several oxidative and conjugative processes. In the present study, the identity of human uridine 5'-diphosphoglucuronyltransferases (UGTs) capable of catalyzing the 25OHD3 glucuronidation reaction was investigated. Two isozymes, UGT1A4 and UGT1A3, were identified as the principal catalysts of 25OHD3 glucuronidation in human liver. Three 25OHD3 monoglucuronides (25OHD3-25-glucuronide, 25OHD3-3-glucuronide, and 5,6-trans-25OHD3-25-glucuronide) were generated by recombinant UGT1A4/UGT1A3, human liver microsomes, and human hepatocytes. The kinetics of 25OHD3 glucuronide formation in all systems tested conformed to the Michaelis-Menten model. An association between the UGT1A4*3 (Leu48Val) gene polymorphism with the rates of glucuronide formation was also investigated using human liver microsomes isolated from 80 genotyped livers. A variant allele dose effect was observed: the homozygous UGT1A4*3 livers (GG) had the highest glucuronidation activity, whereas the wild type (TT) had the lowest activity. Induction of UGT1A4 and UGT1A3 gene expression was also determined in human hepatocytes treated with pregnane X receptor/constitutive androstane receptor agonists, such as rifampin, carbamazepine, and phenobarbital. Although UGT mRNA levels were increased significantly by all of the known pregnane X receptor/constitutive androstane receptor agonists tested, rifampin, the most potent of the inducers, significantly induced total 25OHD3 glucuronide formation activity in human hepatocytes measured after 2, but not 4 and 24 hours, of incubation. Finally, the presence of 25OHD3-3-glucuronide in both human plasma and bile was confirmed, suggesting that the glucuronidation pathway might be physiologically relevant and contribute to vitamin D homeostasis in humans.
Product Description
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, analytical standard
Sigma-Aldrich
3-epi-25-Hydroxyvitamin D3, 98% (CP)
Sigma-Aldrich
Levetiracetam, ≥98% (HPLC)
Sigma-Aldrich
4-Phenyl-1,2,4-triazoline-3,5-dione, 97%
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Hyperforin, ≥85% (HPLC)
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
USP
Levetiracetam, United States Pharmacopeia (USP) Reference Standard
Supelco
Levetiracetam, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Levetiracetam, European Pharmacopoeia (EP) Reference Standard