Merck
  • A role for DNA polymerase θ in promoting replication through oxidative DNA lesion, thymine glycol, in human cells.
A role for DNA polymerase θ in promoting replication through oxidative DNA lesion, thymine glycol, in human cells.
The Journal of biological chemistry (2014-03-22)
Jung-Hoon Yoon, Jayati Roy Choudhury, Jeseong Park, Satya Prakash, Louise Prakash
ABSTRACT
The biological functions of human DNA polymerase (pol) θ, an A family polymerase, have remained poorly defined. Here we identify a role of polθ in translesion synthesis (TLS) in human cells. We show that TLS through the thymine glycol (TG) lesion, the most common oxidation product of thymine, occurs via two alternative pathways, in one of which, polymerases κ and ζ function together and mediate error-free TLS, whereas in the other, polθ functions in an error-prone manner. Human polθ is comprised of an N-terminal ATPase/helicase domain, a large central domain, and a C-terminal polymerase domain; however, we find that only the C-terminal polymerase domain is required for TLS opposite TG in human cells. In contrast to TLS mediated by polκ and polζ, in which polζ would elongate the chain from the TG:A base pair formed by polκ action, the ability of polθ alone to carry out the nucleotide insertion step, as well as the subsequent extension step that presents a considerable impediment due to displacement of the 5' template base, suggests that the polθ active site can accommodate highly distorting DNA lesions.
Sigma-Aldrich
DNA Polymerase I, Klenow Fragment from Escherichia coli, buffered aqueous glycerol solution
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Supelco
Thymine(Zidovudine Related Compound C), Pharmaceutical Secondary Standard; Certified Reference Material