Merck
  • Structural modifications and tissue response after standard epi-off and iontophoretic corneal crosslinking with different irradiation procedures.
Structural modifications and tissue response after standard epi-off and iontophoretic corneal crosslinking with different irradiation procedures.
Investigative ophthalmology & visual science (2014-03-29)
Leonardo Mastropasqua, Manuela Lanzini, Claudia Curcio, Roberta Calienno, Rodolfo Mastropasqua, Martina Colasante, Alessandra Mastropasqua, Mario Nubile
ABSTRACT
The aim of this study is to investigate modifications in human cadaver corneas after different crosslinking procedures, including standard epi-off treatment, iontophoresis imbibition, and different exposure to ultraviolet A (UVA) sources (30 minutes at 3 mW and 9 minutes at 10 mW). A total of 12 human cadaver corneas was examined and divided as follows: 3 served as control (group 1), 3 were treated with a standard epi-off procedure (group 2), 6 underwent iontophoresis imbibition for 5 minutes, and then 3 were irradiated for 30 minutes with 3 mW UVA (group 3), and 3 for 9 minutes at 10 mW UVA (group 4). Deformation amplitude index was measured before and after the corneas underwent treatment. After treatment, corneas were prepared for hematoxylin-eosin and immunohistochemistry evaluation. The expression of TUNEL, matrix metalloproteinase-1 (MMP-1), collagen type I, and CD34 was investigate in all samples. The deformation amplitude index decreased in all groups, in particular in group 4, indicating an improvement of corneal biomechanical properties. Immunohistochemical staining showed a significant stromal alteration in group 2, mild damage in group 3, and no modifications in corneal morphology in group 4. The TUNEL (P < 0.001) and MMP-1 (P = 0.002) positivity was more evident in group 4. Collagen type I positivity significantly increased in groups 3 (P = 0.002) and 4 (P = 0.002). The CD34 expression was more evident in groups 2 (P = 0.003) and 3 (P = 0.003). Iontophoresis imbibition followed by UVA irradiation for 9 minutes at 10 mW determined less tissue damage and better stromal remodeling.
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Anti-Collagen I α2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder