Merck
  • Evaluating in vivo delivery of riboflavin with coulomb-controlled iontophoresis for corneal collagen cross-linking: a pilot study.
Evaluating in vivo delivery of riboflavin with coulomb-controlled iontophoresis for corneal collagen cross-linking: a pilot study.
Investigative ophthalmology & visual science (2014-03-29)
Alejandro Arboleda, Laura Kowalczuk, Michèle Savoldelli, Christophe Klein, Sophia Ladraa, Marie-Christine Naud, Mariela C Aguilar, Jean-Marie Parel, Francine Behar-Cohen
ABSTRACT
To evaluate the efficacy of coulomb-controlled iontophoresis (CCI) for delivery of riboflavin prior to corneal collagen cross-linking (CXL). The eyes of 20 8-week-old Lewis rats, subject to epithelium-ON (epi-ON, n = 20 eyes) or epithelium-OFF (epi-OFF, n = 20 eyes) conditions, were used to evaluate the in vivo delivery of two riboflavin solutions: 0.1% riboflavin-20% dextran T500 solution (riboflavin-dextran) and 0.1% riboflavin 5'-phosphate (riboflavin-phosphate). After systemic intramuscular anesthesia, 0.25 mL of the photosensitizing agent was delivered by either instillation or CCI (2.11 mA/cm(2) for 4 or 10 minutes) into either epithelial condition. The CCI probe on the eye without current served as control. Confocal microscopy of flat-mounted corneas was used to analyze intracorneal penetration and fluorometry was used to quantify riboflavin in the aqueous within 30 minutes of treatment. Instillation and CCI allowed for uniform delivery of riboflavin-dextran throughout the stroma after epithelial debridement. Transepithelial delivery of riboflavin-dextran was not efficacious. Riboflavin-phosphate was successfully delivered in both epithelium conditions. Complete saturation of the cornea was achieved using CCI after removing the epithelium, the epi-ON case allowed for limited diffusion. Increasing the time from 4 to 10 minutes greatly increased the amount of riboflavin detected in the cornea and aqueous humor. Coulomb-controlled iontophoresis is an effective technique for transepithelial delivery of riboflavin-phosphate into the cornea. This drug delivery method would allow clinicians to significantly shorten the time required for the CXL procedure, with or without epithelial debridement. Whether efficient crosslinking can be achieved through an intact epithelium remains to be demonstrated.
Product Description
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Riboflavin, European Pharmacopoeia (EP) Reference Standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Riboflavin 5′-phosphate sodium salt hydrate, meets USP testing specifications
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance