Merck
Exercise stays central to treatment of osteoarthritis in latest advice.
Nursing older people (2014-03-29)
Lisa Berry
Product Description
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard