Merck
  • Degradation of phenol via wet-air oxidation over CuO/CeO2-ZrO2 nanocatalyst synthesized employing ultrasound energy: physicochemical characterization and catalytic performance.
Degradation of phenol via wet-air oxidation over CuO/CeO2-ZrO2 nanocatalyst synthesized employing ultrasound energy: physicochemical characterization and catalytic performance.
Environmental technology (2014-04-08)
Mohsen Parvas, Mohammad Haghighi, Somaiyeh Allahyari
ABSTRACT
Catalytic wet air oxidation (CWAO) of phenol was carried out under atmospheric pressure of oxygen at 160 degrees C in a stirred batch reactor over copper catalysts supported by CeO2-ZrO2. The copper with different loadings were impregnated over the composite support by a sonication process. The catalysts were characterized by X-ray diffraction (XRD), field emission scanning electron microscopy (FESEM), energy dispersive X-ray analysis (EDX), Brunauer-Emmett-Teller (BET) specific surface area and Fourier-transformed infrared analyses. Characteristic peaks attributed to copper were not found in XRD patterns even at high loadings, but based on EDX results, the existence of copper particles was confirmed. It means that sonochemical synthesis method even at high loadings produced small copper particles with low crystallinity and excellent dispersion over the CeO2-ZrO2 composite. FESEM micrographs indicated just slight enhancement in particle size at high loadings of Cu. Blank CWAO experiments illustrated low conversion of phenol using bare CeO2-ZrO2 support. Although some agglomeration of particles was found at high loadings of copper but owning to the fact that almost all ZrO2 particles incorporated into the CeO2 lattice at high contents of Cu, catalyst activity not only did not decrease but also the phenol conversion reached to the higher values. The optimal catalyst loading for phenol degradation was found to be 9 g/l. Complete conversion of phenol was achieved using CuO/CeO2-ZrO2 in 9 g/l catalyst loading with initial phenol concentration of 1000 ppm after 3 h of reaction.
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Copper, insulated wire, 20m, conductor diameter 0.5mm, insulation thickness 0.06mm, polyimide insulation
Zirconium, foil, 15mm disks, thickness 0.5mm, annealed, 99.2%
Copper, insulated wire, 2m, conductor diameter 0.45mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.01mm, polyimide insulation, 99.99%
Copper, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Copper, insulated wire, 5m, conductor diameter 0.45mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper, insulated wire, 5m, conductor diameter 0.1mm, insulation thickness 0.015mm, 10-30 micron polyesterimide insulation, 99.9%
Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%
Copper, insulated wire, 2m, conductor diameter 1.0mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper, insulated wire, 200m, conductor diameter 0.5mm, insulation thickness 0.06mm, polyimide insulation
Copper, rod, 100mm, diameter 4.8mm, hard, 99.9%
Copper, insulated wire, 20m, conductor diameter 0.45mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper, insulated wire, 10m, conductor diameter 0.5mm, insulation thickness 0.06mm, polyimide insulation