Merck
  • Determination of vanadium in groundwater samples with an improved kinetic spectrophotometric method.
Determination of vanadium in groundwater samples with an improved kinetic spectrophotometric method.
Environmental technology (2014-04-08)
Esra Bağda
ABSTRACT
A kinetic catalytic method has been developed for the determination of vanadium based on its catalytic effect on the redox reaction of azorubin S and bromate in the presence of a sulphuric and nitric acid mixture. The reaction is monitored spectrophotometrically by measuring the decrease in absorbance of the reaction mixture at 515 nm. The fixed-time method was used for 0.5-5 min. Optimization of the reaction conditions regarding concentrations of acids, dye, oxidant, masking agent, etc. was investigated. The rate of decrease in absorbance of azorubin S was proportional to the concentration of vanadium in the range of 2.0-1.05 x 10(3) ng mL(-1). 3Sb/m was 0.0129 ng mL(-1) and 10 Sb/m was 0.0432 ng mL(-1). The catalytic method based on the oxidation reaction of azorubin S and bromate shows a good selectivity for vanadium over a wide variety of interference cations and anions. The proposed method was successfully applied to the determination of vanadium in groundwater samples and spiked-water samples.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carmoisine, analytical standard
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Chromotrope FB, Dye content 50 %
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%