Merck
  • Kinetics of struvite precipitation in synthetic biologically treated swine wastewaters.
Kinetics of struvite precipitation in synthetic biologically treated swine wastewaters.
Environmental technology (2014-04-08)
Aurélie Capdevielle, Eva Sýkorová, Fabrice Béline, Marie-Line Daumer
ABSTRACT
An experimental design was set up to understand the influence of five process parameters on the kinetics of struvite precipitation in synthetic swine wastewaters. The responses studied were the kinetics of phosphorus (P) removal, the struvite precipitation rate and the dissolution rate of amorphous calcium phosphates (ACP). The kinetic study showed that the P-removal was complete in less than 1 h and was influenced positively by the added MgO. The precipitation of struvite with MgO was confirmed to follow a first-order kinetic. This study showed that ACP co-precipitated with struvite during the first 30 min. Afterwards, ACP dissolved to maintain the phosphates balance limiting the struvite growth. An initial Mg:Ca > 1.5 induced a complete dissolution of ACP in 1 h. Another experiment was conducted and it validated the results of the statistical model. This experiment also determined that 7-10 h was the best time to recover large crystals. After 10 h, the crystals were broken by stirring.
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh