Merck
  • Efficient removal of crystal violet dye from aqueous solutions by vitreous tuff mineral.
Efficient removal of crystal violet dye from aqueous solutions by vitreous tuff mineral.
Environmental technology (2014-04-08)
A Blanco-Flores, A Colín-Cruz, E Gutiérrez-Segura, V Sánchez-Mendieta, D A Solís-Casados, M A Garrudo-Guirado, R Batista-González
ABSTRACT
Textural, structural and morphological characteristics of the vitreous tuff were determined by means of several physicochemical techniques. The nitrogen adsorption isotherm at 77 K was fitted with the Brunnauer-Emmet-Teller model and together with the results of the average pore distribution showed a mesoporous material. Samples of vitreous tuff were used as adsorbent to study the removal of crystal violet from aqueous solution. The presence of -OH moieties in the material seems to be responsible for the removal of the dye showing that vitreous tuff can be used as an organic dye adsorbent material. The pseudo-second-order model was the best fit model for describing the sorption process of crystal violet; intraparticle diffusion being the controlling step in the process. The experimental adsorption isotherm was fitted with Langmuir, Freundlich and Langmuir-Freundlich models, showing better correlation with the second one. The adsorption capacity was 170.01 mg/g, being among the highest compared with other inorganic and organic common sorbent materials. The design of single stage of the adsorber can predict the behaviour to potential scale up. This mineral has a very good potential as an adsorbent material for organic dyes.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, suitable for microscopy
Methylrosanilinium chloride, European Pharmacopoeia (EP) Reference Standard
Methylrosanilinium for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, S. No.: 785