Merck
  • Conditions and constraints for astrocyte calcium signaling in the hippocampal mossy fiber pathway.
Conditions and constraints for astrocyte calcium signaling in the hippocampal mossy fiber pathway.
Neuron (2014-04-20)
Martin D Haustein, Sebastian Kracun, Xiao-Hong Lu, Tiffany Shih, Olan Jackson-Weaver, Xiaoping Tong, Ji Xu, X William Yang, Thomas J O'Dell, Jonathan S Marvin, Mark H Ellisman, Eric A Bushong, Loren L Looger, Baljit S Khakh
ABSTRACT
The spatiotemporal activities of astrocyte Ca²⁺ signaling in mature neuronal circuits remain unclear. We used genetically encoded Ca²⁺ and glutamate indicators as well as pharmacogenetic and electrical control of neurotransmitter release to explore astrocyte activity in the hippocampal mossy fiber pathway. Our data revealed numerous localized, spontaneous Ca²⁺ signals in astrocyte branches and territories, but these were not driven by neuronal activity or glutamate. Moreover, evoked astrocyte Ca²⁺ signaling changed linearly with the number of mossy fiber action potentials. Under these settings, astrocyte responses were global, suppressed by neurotransmitter clearance, and mediated by glutamate and GABA. Thus, astrocyte engagement in the fully developed mossy fiber pathway was slow and territorial, contrary to that frequently proposed for astrocytes within microcircuits. We show that astrocyte Ca²⁺ signaling functionally segregates large volumes of neuropil and that these transients are not suited for responding to, or regulating, single synapses in the mossy fiber pathway.
Product Description
Supelco
γ-Aminobutyric acid, analytical standard
Sigma-Aldrich
L-Glutamic acid hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)