Merck
  • [A retrospective study of tegafur/uracil compared with cyclophosphamide, methotrexate, and fluorouracil as adjuvant chemotherapy in patients with node-negative, triple-negative breast cancer].
[A retrospective study of tegafur/uracil compared with cyclophosphamide, methotrexate, and fluorouracil as adjuvant chemotherapy in patients with node-negative, triple-negative breast cancer].
Gan to kagaku ryoho. Cancer & chemotherapy (2014-04-20)
Koya Fushimi, Naoto Tanabe, Makoto Takami
ABSTRACT
Japanese clinical trials of a tegafur/uracil(UFT)-based postoperative chemotherapy regimen compared with cyclophosphamide, methotrexate, and fluorouracil(CMF)treatment have shown that UFT is not inferior to CMF for the treatment of hormone receptor-positive breast cancer patients. However, the usefulness of UFT for hormone receptor-negative breast cancer, including the triple-negative subtype(hormone receptor-negative, human epidermal growth factor receptor 2 [HER2]-negative), is unknown. The aim of this retrospective study was to examine the effectiveness of postoperative, adjuvant UFT compared to CMF when these regimens were given to women with node-negative, triple-negative breast cancer. We analyzed 50 women with node-negative, triple-negative breast cancer who were treated with six cycles of CMF(n=33) or two years of UFT(n=17)after surgery between January 2000 and December 2010. Although no significant differences were observed in relapse-free survival or overall survival in all patients, subset analysis of tumors of larger size(t2), higher nuclear grade(NG3), and with positivity for lymphovascular invasion showed that UFT was inferior to CMF. These results encourage caution regarding the choice of adjuvant UFT for node-negative, triple-negative breast cancer.
Sigma-Aldrich
Tegafur, ≥98% (HPLC), powder
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
5-Fluorouracil, analytical standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Methotrexate