Merck
  • [Effect of capecitabine therapy on the blood levels of antiepileptic drugs - report of two cases].
[Effect of capecitabine therapy on the blood levels of antiepileptic drugs - report of two cases].
Gan to kagaku ryoho. Cancer & chemotherapy (2014-04-20)
Hiroyuki Tanaka, Hiromi Jotoku, Masahiko Takasaki, Yukihiro Ibayashi, Kenichi Watanabe, Masato Takahashi
ABSTRACT
We report the cases of 2 breast cancer patients who received capecitabine(CAP)and concomitant anticonvulsant therapy with either phenytoin(PHT)or valproate(VPA)for brain metastasis. The effect of CAP on the blood levels of the 2 anticonvulsants was different and it depended on the variation in metabolism of each drug. Case 1 involved a 59-year-old woman with recurrent breast cancer. After radiation therapy for brain metastases, the patient received PHT(400mg/day)to prevent convulsions. After 5 days of PHT administration, CAP therapy was initiated, and her blood PHT levels increased to 33.8 mg/mL. Although the PHT dose was reduced to 300mg/day, the blood PHT levels markedly increased to 45.5 mg/mL 7 days after the withdrawal of CAP. Case 2 involved a 60-year-old woman with breast cancer who underwent surgery for brain metastases and subsequently received controlled-release VPA tablets(400mg/day). No remarkable change was observed in her blood VPA levels during CAP treatment or after CAP withdrawal(the blood VAP level after 7 days of treatment was, 78.4 mg/mL; after 14 days of treatment, 73.2 mg/mL; and 7 days after withdrawal, 79.7 mg/mL). CAP has been reported to inhibit nucleic acid synthesis and/or folic acid activity rather than cytochrome P450(CYP)directly. CAP had a significant effect on the blood levels of PHT, which is metabolized via the CYP pathway. However, VPA levels remained unchanged because VPA metabolism involves other pathways, such as the beta-oxidation and conjugation pathways.
Product Description
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
2-Propylpentanoic acid
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Capecitabine, ≥98% (HPLC)
Supelco
5-Fluorouracil, analytical standard
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Capecitabine, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Phenytoin, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenytoin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
Supelco
Phenytoin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Phenytoin for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Phenytoin, United States Pharmacopeia (USP) Reference Standard