Merck
  • BARP suppresses voltage-gated calcium channel activity and Ca2+-evoked exocytosis.
BARP suppresses voltage-gated calcium channel activity and Ca2+-evoked exocytosis.
The Journal of cell biology (2014-04-23)
Pascal Béguin, Kazuaki Nagashima, Ramasubbu N Mahalakshmi, Réjan Vigot, Atsuko Matsunaga, Takafumi Miki, Mei Yong Ng, Yu Jin Alvin Ng, Chiaw Hwee Lim, Hock Soon Tay, Le-Ann Hwang, Dmitri Firsov, Bor Luen Tang, Nobuya Inagaki, Yasuo Mori, Susumu Seino, Thomas Launey, Walter Hunziker
ABSTRACT
Voltage-gated calcium channels (VGCCs) are key regulators of cell signaling and Ca(2+)-dependent release of neurotransmitters and hormones. Understanding the mechanisms that inactivate VGCCs to prevent intracellular Ca(2+) overload and govern their specific subcellular localization is of critical importance. We report the identification and functional characterization of VGCC β-anchoring and -regulatory protein (BARP), a previously uncharacterized integral membrane glycoprotein expressed in neuroendocrine cells and neurons. BARP interacts via two cytosolic domains (I and II) with all Cavβ subunit isoforms, affecting their subcellular localization and suppressing VGCC activity. Domain I interacts at the α1 interaction domain-binding pocket in Cavβ and interferes with the association between Cavβ and Cavα1. In the absence of domain I binding, BARP can form a ternary complex with Cavα1 and Cavβ via domain II. BARP does not affect cell surface expression of Cavα1 but inhibits Ca(2+) channel activity at the plasma membrane, resulting in the inhibition of Ca(2+)-evoked exocytosis. Thus, BARP can modulate the localization of Cavβ and its association with the Cavα1 subunit to negatively regulate VGCC activity.
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%