Merck
  • Comorbidity is an independent prognostic factor in patients with advanced-stage diffuse large B-cell lymphoma treated with R-CHOP: a population-based cohort study.
Comorbidity is an independent prognostic factor in patients with advanced-stage diffuse large B-cell lymphoma treated with R-CHOP: a population-based cohort study.
British journal of haematology (2014-04-24)
Andre Wieringa, Karin Boslooper, Mels Hoogendoorn, Peter Joosten, Tim Beerden, Huib Storm, Robby E Kibbelaar, Gerrit J Veldhuis, Harmen van Kamp, Bastiaan van Rees, Hanneke C Kluin-Nelemans, Nic J G M Veeger, Eric N van Roon
ABSTRACT
An observational population-based cohort study was performed to investigate the role of comorbidity on outcome and treatment-related toxicity in patients with newly diagnosed advanced-stage diffuse large B-cell lymphoma (DLBCL) treated with R-CHOP (rituximab, cyclophosphamide, doxorubicin, vincristine, prednisone). Data for the clinical characteristics of 154 patients (median age 69 years), including Charlson Comorbidity Index (CCI), treatment, toxicity and outcome were evaluated. Forty-five percent of the patients had an International Prognistic index ≥3 and 16% had a CCI ≥2. The planned R-CHOP schedule was completed by 84% and 75% reached complete remission (CR). In those with CCI ≥2, 67% completed treatment with 46% CR. In patients with a CCI <2, overall survival (OS) after 1, 2 and 5 years was 84%, 79% and 65% respectively and it was 64%, 48% and 48% for those with CCI ≥2. Grade III/IV toxicity was documented in 53%, most frequently febrile neutropenia (27%) and infections (23%). In multivariate analysis CCI ≥2 and IPI ≥3 were independent risk indicators for OS and grade III/IV toxicity. In conclusion, comorbidity is an independent risk indicator for worse OS in patients with advanced DLBCL treated with R-CHOP by interference with intensive treatment schedules and more grade III/IV toxicity. Future studies are warranted to determine the optimal treatment approach in patients with significant comorbidities.
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard