Merck
  • Succinimide and saccharin-based enzyme-activated inhibitors of serine proteases.
Succinimide and saccharin-based enzyme-activated inhibitors of serine proteases.
Current pharmaceutical design (1999-07-03)
D C Martyn, M J Moore, A D Abell
ABSTRACT
The inhibition of human leukocyte elastase (HLE), and other serine proteases, by succinimide and saccharin-based compounds is reviewed. The succinimide compounds are unique in that the inactivating species is generated within the enzyme active site via a molecular rearrangement. The related saccharin derivatives also inactivate serine proteases by an enzyme-activated mechanism. Those factors effecting the potency, selectivity and stability of these important classes of inhibitor are discussed.
Sigma-Aldrich
Succinimide
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Saccharin, ≥98%
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Saccharin, European Pharmacopoeia (EP) Reference Standard
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard