Merck
A review of the developmental and reproductive toxicity of styrene.
Regulatory toxicology and pharmacology : RTP (2001-02-13)
N A Brown, J C Lamb, S M Brown, B H Neal
ABSTRACT
The reproductive and developmental toxicity of styrene has been studied in animals and humans. The animal studies on styrene have diverse study designs and conclusions. Developmental or reproductive toxicity studies have been conducted in rats, mice, rabbits, and hamsters. In most cases, high doses are required to elicit effects, and the effects are not unique to reproduction or development. In a number of the reports, either the experimental designs are limited or the descriptions of the designs and the endpoints measured are insufficient to draw conclusions about the toxicity of styrene. The more complete and better-reported studies show that styrene does not cause developmental toxicity at dose levels that are not maternally toxic. Some neurochemical or neurobehavioral effects have been reported at high exposures. Styrene does not affect fertility or reproductive function. Considerable animal toxicity data on styrene support the conclusion that styrene is neither an endocrine-active substance nor an endocrine disrupter. Human studies often suffer from either inadequate exposure data or exposure to a wide variety of materials, so that attribution of effects to styrene exposure is impossible. Furthermore, investigators often have failed to account for other exposures in the workplace or for other potentially confounding factors in their studies. Menstrual cycle irregularities and congenital abnormalities were initially reported; however, the better and more recent reports do not show that styrene causes developmental or reproductive effects in humans. Human studies also support the conclusion that styrene is not an endocrine disrupter. Although some study authors have concluded that styrene is either a human or an animal reproductive or developmental toxicant, careful review demonstrates that such conclusions are not justified.
Product Description
Supelco
Styrene, analytical standard
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%