Merck
  • Comparative carcinogenicity of 1,3-butadiene, isoprene, and chloroprene in rats and mice.
Comparative carcinogenicity of 1,3-butadiene, isoprene, and chloroprene in rats and mice.
Chemico-biological interactions (2001-06-09)
R L Melnick, R C Sills
ABSTRACT
1,3-Butadiene, isoprene (2-methyl-1,3-butadiene), and chloroprene (2-chloro-1,3-butadiene) are high-production-volume chemicals used mainly in the manufacture of synthetic rubber. Inhalation studies have demonstrated multiple organ tumorigenic effects with each of these chemicals in mice and rats. Sites of tumor induction by these epoxide-forming chemicals were compared to each other and to ethylene oxide, a chemical classified by the National Toxicology Program (NTP) and by the International Agency for Research on Cancer (IARC) as carcinogenic to humans. For this group of chemicals, there are substantial species differences in sites of neoplasia; neoplasia of the mammary gland is the only common tumorigenic effect in rats and mice. Within each species, there are several common sites of tumor induction; these include the hematopoietic system, circulatory system, lung, liver, forestomach, Harderian gland, and mammary gland in mice, and the mammary gland and possibly the brain, thyroid, testis, and kidney in rats. For studies in which individual animal data were available, mortality-adjusted tumor rates were calculated, and estimates were made of the shape of the exposure-response curves and ED10 values (i.e. exposure concentrations associated with an excess risk of 10% at each tumor site). Most tumorigenic effects reported here were consistent with linear or supralinear models. For chloroprene and butadiene, the most potent response was for the induction of lung neoplasms in female mice, with ED10 values of 0.3 ppm. Based on animal cancer data, isoprene and chloroprene are listed in the NTP's Report on Carcinogens (RoC) as reasonably anticipated to be a human carcinogen. Butadiene is listed in the RoC as known to be a human carcinogen 'based on sufficient evidence of carcinogenicity from studies in humans, including epidemiological and mechanistic information', with support from experimental studies in laboratory animals. Epidemiology data for isoprene and chloroprene are not considered adequate to evaluate the potential carcinogenicity of these agents in humans.
Product Description
SAFC
Isoprene, ≥99%
Sigma-Aldrich
Isoprene, 99%, contains <1000 ppm p-tert-butylcatechol as inhibitor
Supelco
Isoprene, analytical standard
Sigma-Aldrich
1,3-Butadiene solution, 15 wt. % in hexane
Sigma-Aldrich
1,3-Butadiene solution, 20 wt. % in toluene