Merck
  • Dose reduction in skeletal and chest radiography using a large-area flat-panel detector based on amorphous silicon and thallium-doped cesium iodide: technical background, basic image quality parameters, and review of the literature.
Dose reduction in skeletal and chest radiography using a large-area flat-panel detector based on amorphous silicon and thallium-doped cesium iodide: technical background, basic image quality parameters, and review of the literature.
European radiology (2004-02-18)
Markus Völk, Okka W Hamer, Stefan Feuerbach, Michael Strotzer
ABSTRACT
The two most frequently performed diagnostic X-ray examinations are those of the extremities and of the chest. Thus, dose reduction in the field of conventional skeletal and chest radiography is an important issue and there is a need to reduce man-made ionizing radiation. The large-area flat-panel detector based on amorphous silicon and thallium-doped cesium iodide provides a significant reduction of radiation dose in skeletal and chest radiography compared with traditional imaging systems. This article describes the technical background and basic image quality parameters of this 43 x 43-cm digital system, and summarizes the available literature (years 2000-2003) concerning dose reduction in experimental and clinical studies. Due to its high detective quantum efficiency and dynamic range compared with traditional screen-film systems, a dose reduction of up to 50% is possible without loss of image quality.
Sigma-Aldrich
Cesium, ingot, ≥99.95% trace metals basis
Sigma-Aldrich
Thallium, granular, 99.9% trace metals basis
Sigma-Aldrich
Cesium iodide, 99.999% trace metals basis
Sigma-Aldrich
Cesium iodide, 99.9% trace metals basis
Supelco
Cesium iodide, analytical standard, suitable for mass spectrometry (MS)
Sigma-Aldrich
Cesium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis