Merck
Safety assessment of MIBK (methyl isobutyl ketone).
International journal of toxicology (2004-05-28)
Wilbur Johnson
ABSTRACT
MIBK (Methyl Isobutyl Ketone) is an aliphatic ketone that functions as both a denaturant and solvent in cosmetic products. Current use in cosmetic products is very limited, but MIBK is reported to be used in one nail correction pen (volume = 3 ml) at a concentration of 21%. The maximum percutaneous absorption rate in guinea pigs is 1.1 micromol/min/cm2 at 10 to 45 min. Metabolites include 4-hydroxy-4-methyl-2-pentanone (oxidation product) and 4-methyl-2-pentanol (4-MPOL) (reduction product). Values for the serum half-life and total clearance time of MIBK in animals were 66 min and 6 h, respectively. In clinical tests, most of the absorbed MIBK had been eliminated from the body 90 min post exposure. MIBK was not toxic via the oral or dermal route of exposure in acute, short-term, or subchronic animal studies, except that nephrotoxicity was observed in rats dosed with 1 g/kg in a short-term study. MIBK was an ocular and skin irritant in animal tests. Ocular irritation was noted in 12 volunteers exposed to 200 ppm MIBK for 15 min in a clinical test. A depression of the vestibulo-oculomotor reflex was seen with intravenous infusion of MIBK (in an emulsion) at 30 microM/kg/min in female rats. The no-observed-effect level in rats exposed orally to MIBK was 50 mg/kg. Both gross and microscopic evidence of lung damage were reported in acute inhalation toxicity studies in animals. Short-term and subchronic inhalation exposures (as low as 100 ppm) produced effects in the kidney and liver that were species and sex dependent. Dermal doses of 300 or 600 mg/kg for 4 months in rats produced reduced mitotic activity in hair follicles, increased thickness of horny and granular cell layers of the epidermis, a decrease in the number of reactive centers in follicles (spleen), an increase in the number of iron-containing pigments in the area of the red pulp (spleen), and a reduction in the lipid content of the cortical layer of the adrenal glands. Neuropathological changes in the most distal portions of the tibial and ulnar nerves were observed in young adult rats which inhaled 1500 ppm MIBK for up to 5 months. No adverse effects were seen in any other neurological end point by any route of exposure in other studies using rats or other animal species. Clinical tests demonstrated a threshold for MIBK-induced irritation of the lungs at 0.03 to 0.1 mg/L after 1 min of respiration. MIBK was not mutagenic in the Ames test or in a mitotic gene-conversion assay in bacteria. Mammalian mutagenicity test results were also negative in the following assays: mouse lymphoma, unscheduled DNA synthesis, micronucleus, cell transformation, and chromosome damage. MIBK did not induce any treatment-related increases in embryotoxicity or fetal malformations in pregnant Fischer 344 rats or CD-1 mice that inhaled MIBK at concentrations of 300, 1000, or 3000 ppm. There was evidence of treatment-related maternal toxicity only at the highest concentration tested. MIBK applied to the tail of rats daily at doses of 300 or 600 mg/kg for 4 months produced changes in the testes, including a reduction in the number of spermatocytes, spermatids, and spermatozoa. An ongoing carcinogenicity study of MIBK being conducted by the National Toxicology Program will be considered when the results are available. On the basis of the information that is currently available, MIBK is considered safe as used in nail polish removers and as an alcohol denaturant in cosmetic products.
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%, FCC
Supelco
4-Methyl-2-pentanone, analytical standard
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Sigma-Aldrich
4-Methyl-2-pentanone, ACS reagent, ≥98.5%