Merck
  • Influence of triacylglycerol structure of stearic acid-rich fats on postprandial lipaemia.
Influence of triacylglycerol structure of stearic acid-rich fats on postprandial lipaemia.
The Proceedings of the Nutrition Society (2005-06-18)
Sarah E E Berry, Thomas A B Sanders
ABSTRACT
Exaggerated postprandial lipaemia may increase the risk of CHD by contributing to both thrombotic and atherogenic processes. Previous research has focused on the quantity and composition of dietary fat, whereas the effect of triacylglycerol (TAG) structure on postprandial lipaemia and clotting factor VII activity has received little attention. TAG with similar fatty acid composition may have different biochemical and physical properties that are dependent on their TAG structure, and these differences may affect lipid metabolism. Recent findings suggest that differences in the physical properties of stearic acid-rich fats are associated with differences in postprandial lipaemia, and may play an important role in determining their rates of digestion and absorption.
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard