Merck
  • A review of analytical methods for the determination of trichloroethylene and its major metabolites chloral hydrate, trichloroacetic acid and dichloroacetic acid.
A review of analytical methods for the determination of trichloroethylene and its major metabolites chloral hydrate, trichloroacetic acid and dichloroacetic acid.
Biomedical chromatography : BMC (2005-04-14)
Amy D Delinsky, James V Bruckner, Michael G Bartlett
ABSTRACT
Trichloroethylene (TCE) and some of its metabolites are potentially carcinogenic compounds that the general population is commonly exposed to in drinking water. Concentrations of TCE, dichloroacetic acid (DCA) and trichloroacetic acid (TCA) given to laboratory animals in cancer bioassays are high, whereas drinking water levels of the compounds are very low. It is not clear whether the trace amounts of TCE, DCA and TCA in drinking water pose a cancer risk to humans. The accuracy of pharmacokinetic studies relies on the analytical method from which blood and tissue concentration data are obtained. Models that extrapolate cancer risks of TCE and its metabolites from laboratory animals to humans, in turn, rely on the results of pharmacokinetic studies. Therefore, it is essential to have reliable analytical methods for the analysis of TCE and its metabolites. This paper reviews the methods currently in the literature for the analysis of TCE, DCA, TCA and, to a lesser extent, chloral hydrate (CH). Additional aspects of analytical methods such as method validation, species preservation and future directions in the analysis of TCE and its metabolites are also discussed.
Sigma-Aldrich
DCA Deblock (0.36M dichloroacetic acid in toluene)
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Supelco
Dichloroacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Chloral hydrate, ≥99%
Sigma-Aldrich
Chloral hydrate, crystallized, ≥98.0% (T)