- Biology of the molybdenum cofactor.
Biology of the molybdenum cofactor.
Journal of experimental botany (2007-03-14)
Ralf R Mendel
PMID17351249
ABSTRACT
The transition element molybdenum (Mo) is an essential micronutrient for plants where it is needed as a catalytically active metal during enzyme catalysis. Four plant enzymes depend on molybdenum: nitrate reductase, sulphite oxidase, xanthine dehydrogenase, and aldehyde oxidase. However, in order to gain biological activity and fulfil its function in enzymes, molybdenum has to be complexed by a pterin compound thus forming the molybdenum cofactor. In this article, the path of molybdenum from its uptake into the cell, via formation of the molybdenum cofactor and its storage, to the final modification of the molybdenum cofactor and its insertion into apo-metalloenzymes will be reviewed.
Product Description
Molybdenum, wire reel, 50m, diameter 0.25mm, annealed, 99.95%
Molybdenum, foil, 10mm disks, thickness 0.025mm, annealed, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.009mm, 99.9%
Molybdenum, foil, 10m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Molybdenum, foil, 300x300mm, thickness 0.5mm, annealed, 99.9%
Molybdenum, rod, 200mm, diameter 10.0mm, centerless ground, 99.9%
Molybdenum, pellets, 100g, max. size 10mm, 99.9%
Molybdenum, foil, 8mm disks, thickness 0.15mm, annealed, 99.9%
Molybdenum, tube, 100mm, outside diameter 2.0mm, inside diameter 1.3mm, wall thickness 0.35mm, 99.9%
Molybdenum, foil, light tested, 50x50mm, thickness 0.008mm, 99.9%
Molybdenum, foil, light tested, 50x50mm, thickness 0.006mm, 99.9%
Molybdenum, tube, 500mm, outside diameter 1.5mm, inside diameter 0.95mm, wall thickness 0.275mm, 99.9%
Molybdenum, rod, 1000mm, diameter 2.0mm, centerless ground, 99.9%
Molybdenum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Molybdenum, foil, not light tested, 100x100mm, thickness 0.007mm, 99.9%
Molybdenum, foil, 8mm disks, thickness 0.075mm, annealed, 99.9%
Molybdenum, mesh, 150x150mm, nominal aperture 0.44mm, wire diameter 0.07mm, 50 wires/inch, open area 67%, plain weave mesh
Molybdenum, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Molybdenum, foil, not light tested, 50x50mm, thickness 0.002mm, 99.9%
Molybdenum, rod, 200mm, diameter 5.0mm, centerless ground, 99.9%
Molybdenum, rod, 100mm, diameter 3.0mm, centerless ground, 99.9%
Molybdenum, foil, not light tested, 150x150mm, thickness 0.05mm, annealed, 99.9%
Molybdenum, foil, 0.5m coil, thickness 0.0125mm, 99.9%
Molybdenum, rod, 100mm, diameter 15.0mm, centerless ground, 99.9%
Molybdenum, rod, 1000mm, diameter 6.0mm, centerless ground, 99.9%
Molybdenum, wire reel, 50m, diameter 0.025mm, hard, 99.95%
Molybdenum, wire reel, 50m, diameter 0.125mm, annealed, 99.95%