Merck
[Induced plant resistance and salicylic acid: a review].
Prikladnaia biokhimiia i mikrobiologiia (2007-10-13)
N I Vasiukova, O L Ozeretskovskaia
ABSTRACT
Current understanding of the involvement of salicylic acid (SA) in the formation of plant resistance has been reviewed. SA acts as a signal molecule in the SA-dependent pathway. The so-called salicylate burst observed in tissues of plants after stress increases their resistance. The mechanism whereby SA induces plant resistance depends on the ability of this compound to inhibit the enzymes of the antioxidant system of plants, which results in the accumulation of active oxygen species and expression of defense genes.
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Salicylic acid, ≥99%, FG