Merck
Salicylic acid in plant defence--the players and protagonists.
Current opinion in plant biology (2007-10-02)
Gary Loake, Murray Grant
ABSTRACT
Salicylic acid (SA) is synthesised by plants in response to challenge by a diverse range of phytopathogens and is essential to the establishment of both local and systemic-acquired resistance (SAR). SA application induces accumulation of pathogenesis-related (PR) proteins. Mutations leading to either reduced SA production or impaired SA perception enhance susceptibility to avirulent and virulent pathogens. However, our knowledge of the primary signalling components activating SA biosynthesis and linking to PR proteins accumulation is rudimentary. We review progress towards characterising key players (NPR1, MPK4) and processes (methylation, amino acid conjugation, S-nitrosylation) contributing to SA-signalling and perception pathways. Further, we examine emerging data on how pathogens have evolved strategies (e.g. ABA modulation and coronatine production) to suppress SA-mediated plant defence.
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Salicylic acid, European Pharmacopoeia (EP) Reference Standard