Merck
Metabolic pathways of T-2 toxin.
Current drug metabolism (2008-01-29)
Vlastimil Dohnal, Alena Jezkova, Daniel Jun, Kamil Kuca
ABSTRACT
Among the naturally-occurring trichothecenes found in food and feed, T-2 toxin is the most potent and toxic mycotoxin. After ingestion of T-2 toxin into the organism, it is processed and eliminated. Some metabolites of this trichothecene are equally toxic or slightly more toxic than T-2 itself, and therefore, the metabolic fate of T-2 toxin has been of great concern. The main reactions in trichothecene metabolism are hydrolysis, hydroxylation and deep oxidation. Typical metabolites of T-2 toxin in an organism are HT-2 toxin, T-2-triol, T-2-tetraol, 3'-hydroxy-T-2, and 3'-hydroxy-HT-2 toxin. There are significant differences in the metabolic pathways of T-2 toxin between ruminants and non-ruminants. Ruminants have been more resistant to the adverse effects of T-2 toxin due to microbial degradation within rumen microorganisms. Some plant species are resistant to T-2 toxin, while others are capable of its intake and metabolisation.
Product Description
Supelco
T2-Toxin solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)