Merck
Sodium bicarbonate: basically useless therapy.
Pediatrics (2008-10-03)
Judy L Aschner, Ronald L Poland
ABSTRACT
Common clinical practices often are unsupported by experimental evidence. One example is the administration of sodium bicarbonate to neonates. Despite a long history of widespread use, objective evidence that administration of sodium bicarbonate improves outcomes for patients in cardiopulmonary arrest or with metabolic acidosis is lacking. Indeed, there is evidence that this therapy is detrimental. This review examines the history of sodium bicarbonate use in neonatology and the evidence that refutes the clinical practice of administering sodium bicarbonate during cardiopulmonary resuscitation or to treat metabolic acidosis in the NICU.
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C