Merck
Ergogenic effects of sodium bicarbonate.
Current sports medicine reports (2008-07-09)
Lars R McNaughton, Jason Siegler, Adrian Midgley
ABSTRACT
Athletes use many different strategies to enhance their performance, including clothing and footwear, training regimes, diets, and ergogenic aids. The use of ergogenic aids is believed to be widespread, with a variety of legal as well as illegal substances being used previously and currently. Among the more popular ergogenic aids is the use of sodium bicarbonate or sodium citrate, collectively recognized as "buffers." These substances potentially provide the body with added resistance against fatigue caused by deleterious changes in acid-base balance brought about by a variety of exercise modes and durations. The popularity of buffering has generated a plethora of research dating back to the 1930s, which continues to date. The issues surrounding buffering revolve around the dosage size, timing of ingestion, and the type of exercise to benefit from the use of buffers. We hope this review addresses these pertinent issues.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder