Merck
Encapsulated mesalamine granules (Apriso) for ulcerative colitis.
The Medical letter on drugs and therapeutics (2009-05-19)
ABSTRACT
Apriso (Salix) is a new formulation of mesalamine (5-aminosalicylic acid; 5-ASA) approved by the FDA for maintenance of remission in mild to moderate ulcerative colitis (UC). Mesalamine is a locally acting antiinflammatory agent that is widely used both to maintain and induce remission in inflammatory bowel disease. Various mesalamine formulations have been developed to target drug delivery to areas of the small intestine and colon. Most of these agents require frequent dosing and have a high pill burden. The newest products--Lialda, introduced in 2007, and now Apriso--can be dosed once daily.
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Mesalazine, European Pharmacopoeia (EP) Reference Standard
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard