Merck
  • Bioconjugation with stable luminescent lanthanide(III) chelates comprising pyridine subunits.
Bioconjugation with stable luminescent lanthanide(III) chelates comprising pyridine subunits.
Bioconjugate chemistry (2008-12-17)
Jari Hovinen, Pamela M Guy
ABSTRACT
The use of long-lifetime emitting lanthanide(III) chelate labels or probes together with time-resolved fluorometry in detection provides a method to generate sensitive bioaffinity assays. However, the use of stable chelates demands very complicated optimization of the chelate structure. A great number of chelates have been synthesized, but usually, only the most prominent structures were then converted to corresponding biomolecule labeling reactants. This review covers the syntheses of luminescent lanthanide chelates comprising a pyridine subunit that allow solution and solid-phase bioconjugation.
Product Description
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%