Merck
Review of phenol treatment in sacrococcygeal pilonidal disease.
Techniques in coloproctology (2009-08-06)
Cuneyt Kayaalp, Cemalettin Aydin
ABSTRACT
Investigating minimally invasive methods for treatment of pilonidal disease, we reviewed the techniques and the results of phenol treatment of pilonidal disease in the medical literature. A Pubmed search for all English language written papers and abstracts published between January 1964 and September 2007. Mean time to return to work is 2.3 +/- 3.8 days and mean healing time is 20 +/- 14 days. Overall success rate is 87 +/- 10% with a mean follow-up of 2.0 +/- 1.1 years. The most common postoperative complications after phenolization are development of abscesses and cellulites. The incidence of morbidities was mean 8.9 +/- 4.7%. Success rate looks better particularly in the cases that have 1-3 sinus orifices and comparable with the surgical methods. Though healing time of the wound is long, the procedure apparently lessens the time off work. Although results of this review indicate that phenol treatment may be beneficial for pilonidal disease, the lack of randomized studies results in only weak evidence. There is also the need for longer-term follow-up data.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%