Merck
  • The ADVANCE trial: clarifying the role of perindopril/indapamide fixed-dose combination in the reduction of cardiovascular and renal events in patients with diabetes mellitus.
The ADVANCE trial: clarifying the role of perindopril/indapamide fixed-dose combination in the reduction of cardiovascular and renal events in patients with diabetes mellitus.
American journal of cardiovascular drugs : drugs, devices, and other interventions (2009-10-02)
Bernard Waeber, Alejandro de la Sierra, Luis M Ruilope
ABSTRACT
Patients with type 2 diabetes mellitus exhibit a marked increase in cardiovascular and renal risk. A number of interventional trials have shown that these patients benefit greatly from aggressive BP lowering, especially when the drug regimen comprises an inhibitor of the renin-angiotensin system. The results of the placebo-controlled ADVANCE (Action in Diabetes and Vascular disease: PreterAx and DiamicroN MR Controlled Evaluation) trial, conducted in patients with type 2 diabetes, are exemplary in this respect. The systematic use of a fixed-dose combination containing the ACE inhibitor perindopril and the diuretic indapamide afforded substantial protection against cardiovascular mortality and myocardial infarction, while providing important renoprotection, reducing the development of micro- and macroalbuminuria, and allowing regression of nephropathy. The beneficial effects were obtained regardless of baseline BP and whether or not the patients were receiving antihypertensive therapy.
Product Description
Supelco
Indapamide, analytical standard, for drug analysis
Indapamide, European Pharmacopoeia (EP) Reference Standard
USP
Indapamide, United States Pharmacopeia (USP) Reference Standard