Merck
Fructose-sorbitol malabsorption.
Current gastroenterology reports (2009-09-22)
Fernando Fernández-Bañares, Maria Esteve, Josep M Viver
ABSTRACT
Important dietary carbohydrates such as fructose and sorbitol are incompletely absorbed in the normal small intestine. This malabsorption is sometimes associated with abdominal complaints and diarrhea development, symptoms indistinguishable from those of functional bowel disease. Recently, polymerized forms of fructose (fructans) also were implicated in symptom production in patients with irritable bowel syndrome (IBS). Evidence from uncontrolled and controlled challenge studies suggests that malabsorbed sugars (fructose, sorbitol, lactose) and fructans may act as dietary triggers for clinical symptoms suggestive of IBS. Further placebo-controlled studies are needed to obtain definite conclusions about the role of dietary sugar malabsorption in functional bowel disease.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard