Merck
  • Physiological and performance effects of glycerol hyperhydration and rehydration.
Physiological and performance effects of glycerol hyperhydration and rehydration.
Nutrition reviews (2009-11-28)
Simon P van Rosendal, Mark A Osborne, Robert G Fassett, Jeff S Coombes
ABSTRACT
Studies have shown that beverages containing glycerol can enhance and maintain hydration status and may improve endurance exercise performance by attenuating adverse physiological changes associated with dehydration. Improvements to performance include increased endurance time to exhaustion by up to 24%, or a 5% increase in power or work. However, some studies have found no performance benefits during either prolonged exercise or specific skill and agility tests. In studies that have shown benefits, the improvements have been associated with thermoregulatory and cardiovascular changes. These include increased plasma volume and sweat rates, as well as reduced core temperature and ratings of perceived exertion. In a very small number of subjects, glycerol consumption has been associated with side-effects including nausea, gastrointestinal discomfort, dizziness, and headaches. In summary, while glycerol and fluid ingestion results in hyperhydration, the documented benefits to exercise performance remain inconsistent.
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material