Merck
A review: advances in microbial remediation of trichloroethylene (TCE).
Journal of environmental sciences (China) (2010-04-20)
Prabhakar Pant, Sudhakar Pant
ABSTRACT
Research works in the recent past have revealed three major biodegradation processes leading to the degradation of trichloroethylene. Reductive dechlorination is an anaerobic process in which chlorinated ethenes are used as electron acceptors. On the other hand, cometabolism requires oxygen for enzymatic degradation of chlorinated ethenes, which however yields no benefit for the bacteria involved. The third process is direct oxidation under aerobic conditions whereby chlorinated ethenes are directly used as electron donors by microorganisms. This review presented the current research trend in understanding biodegradation mechanisms with regard to their field applications. All the techniques used are evaluated, with the focus being on various factors that influence the process and the outcome.
Product Description
USP
Residual Solvent Class 2 - Trichloroethylene, United States Pharmacopeia (USP) Reference Standard
Supelco
Trichloroethylene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%