Merck
  • Plant autophagy puts the brakes on cell death by controlling salicylic acid signaling.
Plant autophagy puts the brakes on cell death by controlling salicylic acid signaling.
Autophagy (2009-12-22)
Kohki Yoshimoto
ABSTRACT
It has long been recognized that autophagy in plants is important for nutrient recycling and plays a critical role in the ability of plants to adapt to environmental extremes such as nutrient deprivation. Recent reverse genetic studies, however, hint at other roles for autophagy, showing that autophagy defects in higher plants result in early senescence and excessive immunity-related programmed cell death (PCD), irrespective of nutrient conditions. Until now, the mechanisms by which cells die in the absence of autophagy were unclear. In our study, using biochemical, pharmacological and genetic approaches, we reveal that excessive salicylic acid (SA) signaling is a major factor in autophagy-defective plant-dependent cell death and that the SA signal can induce autophagy. These findings suggest a novel physiological function for plant autophagy that operates via a negative feedback loop to modulate proper SA signaling.
Product Description
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard