Merck
  • [Calcipotriol plus betamethasone dipropionate gel in the treatment of scalp psoriasis. A review].
[Calcipotriol plus betamethasone dipropionate gel in the treatment of scalp psoriasis. A review].
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete (2010-07-21)
M A Radtke, K Herberger, T Kornek, M Augustin
ABSTRACT
Scalp psoriasis has a significant psychosocial impact on individuals affecting their quality of life. It occurs in 50-80% of patients with psoriasis and may be the only affected area of the body. Current topical therapies for scalp psoriasis are difficult or unpleasant to apply, resulting in decreased adherence and efficacy. A combination of calcipotriol and betamethasone dipropionate in a gel formulation has been developed and approved in Germany for the treatment of scalp psoriasis. Multiple randomized controlled clinical trials have confirmed the efficacy and safety of the new formulation. After two weeks, 60% of patients showed significant improvement; this climbed to 70%, after eight weeks. It may be safely used for up to 52 weeks. No cases of atrophy, striae, or steroid purpura were noted in two 52-week studies. Although scalp psoriasis can often be adequately treated with topical therapy, there is a need for treatment recommendations and algorithms for severe forms and for patients with extended body involvement, taking the combination of systemic and topical treatment and the presence of comorbidities into account.
Product Description
Betamethasone dipropionate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betamethasone, ≥98%
Sigma-Aldrich
Betamethasone 17,21-dipropionate
Supelco
Betamethasone, VETRANAL®, analytical standard
USP
Betamethasone dipropionate, United States Pharmacopeia (USP) Reference Standard
Betamethasone dipropionate, European Pharmacopoeia (EP) Reference Standard
Betamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Betamethasone Dipropionate, Pharmaceutical Secondary Standard; Certified Reference Material