Merck
Innovative use of topical metronidazole.
Dermatologic clinics (2010-06-01)
Catherine M Zip
ABSTRACT
Metronidazole is a synthetic nitroimidazole derivative with antimicrobial and antiinflammatory properties. It was the first topical therapy approved solely for rosacea and remains a cornerstone of rosacea management. This article reviews the optimal use of topical metronidazole in the treatment of rosacea and other innovative but off-label dermatologic uses reported in the literature.
Product Description
Metronidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material