Merck
  • [The usefulness of nicotinamide in radioiodine therapy in patients with toxic and nontoxic large goitres].
[The usefulness of nicotinamide in radioiodine therapy in patients with toxic and nontoxic large goitres].
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego (2010-08-18)
Małgorzata Natalia Mojsak, Franciszek Rogowski
ABSTRACT
Nicotinamide (niacin) is very useful substance in treatment of many kinds of diseases. For the decades the main indications for application of niacin were lipid disorders. There are studies confirming that niacin increases thyroid radiosensitivity to 1-131. The radioiodine therapy is common method of treatment of hyperthyroidism (Graves' disease, toxic nodular goitre), large euthyroid goitre and differentiated thyroid cancer. In some studies with Wistar rats there is shown that niacin potentiates the effect of 1-131 by increasing thyroid blood flow, what results better effects of radioiodine treatment. Moreover niacin in therapeutic doses decreases serum thyroid hormone levels (mainly total thyroxine and thyroxine-binding globuline). In case of large goitre, when the calculated radioisotope dose exceeds ambulatory limits, the patient must be hospitalized. There are patients with low radioiodine uptake and radiosensitivity of thyroid, radiosensitizers can be utilized for this purpose. Recently obtained results of studies are showing that niacin can be used as radiosensitizer. It is making possible ambulatory treatment of patients with large goitres and low radioiodine sensitive thyroid. The radioiodine therapy with niacin could become shorter, less expensive and more safety for patients.
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard