Merck
Behavioral and neurochemical effects of proline.
Metabolic brain disease (2011-06-07)
Angela T S Wyse, Carlos Alexandre Netto
ABSTRACT
Proline is an amino acid with an essential role for primary metabolism and physiologic functions. Hyperprolinemia results from the deficiency of specific enzymes for proline catabolism, leading to tissue accumulation of this amino acid. Hyperprolinemic patients can present neurological symptoms and brain abnormalities, whose aetiopathogenesis is poorly understood. This review addresses some of the findings obtained, mainly from animal studies, indicating that high proline levels may be associated to neuropathophysiology of some disorders. In this context, it has been suggested that energy metabolism deficit, Na(+),K(+)-ATPase, kinase creatine, oxidative stress, excitotoxicity, lipid content, as well as purinergic and cholinergic systems are involved in the effect of proline on brain damage and spatial memory deficit. The discussion focuses on the relatively low antioxidant defenses of the brain and the vulnerability of neural tissue to reactive species. This offers new perspectives for potential therapeutic strategies for this condition, which may include the early use of appropriate antioxidants as a novel adjuvant therapy, besides the usual treatment based on special diets poor in proline.
Product Description
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
SAFC
L-Proline
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Proline, European Pharmacopoeia (EP) Reference Standard