Merck
  • Clinical features and outcomes of eosinophilic myocarditis patients treated with prednisolone at a single institution over a 27-year period.
Clinical features and outcomes of eosinophilic myocarditis patients treated with prednisolone at a single institution over a 27-year period.
Internal medicine (Tokyo, Japan) (2011-05-03)
Sayaka Kawano, Johji Kato, Noriaki Kawano, Yuki Yoshimura, Hiroyuki Masuyama, Takashi Fukunaga, Yuichiro Sato, Haruhiko Maruyama, Kenroh Mihara, Akira Ueda, Kiyokazu Toyoda, Takuroh Imamura, Kazuo Kitamura
ABSTRACT
Eosinophilic myocarditis is a rare clinical entity characterized by eosinophilia and myocardial inflammation with infiltrating eosinophils. The prognosis of patients with eosinophilic myocarditis is difficult to determine due the disease's rarity and varied causes; consequently, standard treatment has not been established. To elucidate the clinical characteristics and treatment outcome of eosinophilic myocarditis, we retrospectively studied 7 patients fulfilling the criteria of the Japanese Circulation Society for eosinophilic myocarditis from among 64 patients admitted to our institution with eosinophilia over a 27-year period. The patients' ages at diagnosis ranged from 36 to 83 years (median: 52 years). The etiologies of the eosinophilic myocarditis were found to be idiopathic (3 patients), Churg-Strauss syndrome (2 patients), parasitic infection (1 patient) and chronic eosinophilic leukemia (CEL) (1 patient). In addition to treatment for the underlying disease, we also administered prednisolone at a dose appropriate to the disease severity (6 of 7 patients). The patient who was diagnosed with a parasitic infection was treated only with albendazole, because eosinophilic myocarditis was mild. The patient with CEL was positive for the FIP1 L1-PDGFRα fusion gene and was treated with imatinib. Eosinophilic cationic protein was a useful marker for assessing disease activity and treatment efficacy. At the end of the study, of the seven patients treated, six were alive (86%), giving a mean survival time of 37 ± 40 months (mean ± SD). Because eosinophilic myocarditis has various etiologies, it is essential to identify the etiology of the underlying disease. In the majority of eosinophilic myocarditis patients, administration of prednisolone may be an effective therapeutic modality producing a good outcome.
Product Description
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
Prednisolone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prednisolone, ≥99%
Supelco
Prednisolone, VETRANAL®, analytical standard
Prednisolone for peak identification, European Pharmacopoeia (EP) Reference Standard