Merck
  • Recent advances in gemini surfactants: oleic Acid-based gemini surfactants and polymerizable gemini surfactants.
Recent advances in gemini surfactants: oleic Acid-based gemini surfactants and polymerizable gemini surfactants.
Journal of oleo science (2011-03-24)
Kenichi Sakai, Hideki Sakai, Masahiko Abe
ABSTRACT
Gemini surfactants recently developed by our research group are introduced from the standpoints of their syntheses, aqueous solution properties, and potential applications. Two series of gemini surfactants are introduced in this short review, the first of which is the oleic acid-based gemini surfactants, and the second is the polymerizable gemini surfactants. These gemini surfactants have been developed not only as environmentally friendly materials (the use of gemini surfactants enables the reduction of the total consumption of surfactants in chemical products owing to their excellent adsorption and micellization capabilities at low concentrations) but also as functional organic materials.
Product Description
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)