Merck
  • Ascorbic acid: an old player with a broad impact on body physiology including oxidative stress suppression and immunomodulation: a review.
Ascorbic acid: an old player with a broad impact on body physiology including oxidative stress suppression and immunomodulation: a review.
Mini reviews in medicinal chemistry (2011-11-11)
M Pohanka, J Pejchal, S Snopkova, K Havlickova, J Z Karasova, P Bostik, J Pikula
ABSTRACT
Ascorbic acid is a low molecular weight antioxidant well known as anti-scorbut acting vitamin C in humans, primates and guinea pigs. This review summarizes basic data about ascorbic acid in its physiological action point of view. It is divided into biochemistry of ascorbic acid synthesis, mechanism of antioxidant action and participation in anabolism, pharmacokinetics and excretion, exogenous ascorbic acid immunomodulatory effect and participation in infectious diseases, impact on irradiation and intoxication pathogenesis, and supplementary demands. The primary intention was to consider ascorbic acid not only as an antioxidant but also as a chemical compound affecting multiple pathways with a potential beneficial impact in many diseases and processes in human body.
Product Description
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)