Merck
  • Optimization of anti-pseudomonal antibiotics for cystic fibrosis pulmonary exacerbations: I. aztreonam and carbapenems.
Optimization of anti-pseudomonal antibiotics for cystic fibrosis pulmonary exacerbations: I. aztreonam and carbapenems.
Pediatric pulmonology (2012-08-23)
Jeffery T Zobell, David C Young, C Dustin Waters, Chris Stockmann, Krow Ampofo, Catherine M T Sherwin, Michael G Spigarelli
ABSTRACT
Acute pulmonary exacerbations (APE) in cystic fibrosis (CF) are associated with loss of lung function that may require aggressive management with intravenous antibiotics. The aim of this review is to provide an evidence-based summary of pharmacokinetic/pharmacodynamic (PK/PD), tolerability, and efficacy studies utilizing aztreonam and anti-pseudomonal carbapenems (i.e., doripenem, imipenem-cilastatin, and meropenem) in the treatment of an APE, and to identify areas where further study is warranted. The current dosing recommendations in the United States and Europe for aztreonam are lower than the literature supported dosing range of 200-300 mg/kg/day divided every 6 hr, maximum 8-12 g/day. In vitro, PK/PD, and tolerability studies show the potential of doripenem 90 mg/kg/day divided every 8 hr, infused over 4 hr, maximum 6 g/day in the treatment of APE. Imipenem-cilastatin 100 mg/kg/day divided every 6 hr, maximum 4 g/day and meropenem 120 mg/kg/day divided every 8 hr, maximum 6 g/day have been shown to be tolerable and effective in the treatment of APE. With availability issues of new anti-pseudomonal agents and a large percentage of CF patients will not regain their lung function following an APE, we suggest the need to determine optimization of aztreonam and meropenem dosing in CF, as well as to determine the clinical efficacy of doripenem in the treatment of APE. The usefulness of imipenem-cilastatin may be limited due to the rapid development of resistance.
Product Description
USP
Aztreonam, United States Pharmacopeia (USP) Reference Standard
Supelco
Aztreonam, analytical standard