Merck
  • Sodium bicarbonate therapy for the prevention of contrast-induced acute kidney injury – a systematic review and meta-analysis –.
Sodium bicarbonate therapy for the prevention of contrast-induced acute kidney injury – a systematic review and meta-analysis –.
Circulation journal : official journal of the Japanese Circulation Society (2012-09-15)
Jae-Sik Jang, Han-Young Jin, Jeong-Sook Seo, Tae-Hyun Yang, Dae-Kyeong Kim, Tae Hee Kim, Sang-Hwa Urm, Dong-Soo Kim, Dong-Kie Kim, Sang-Hoon Seol, Doo-Il Kim, Kyoung-Im Cho, Bo-Hyun Kim, Yong Hyun Park, Hyung-Gon Je, Jung-Min Ahn, Won-Jang Kim, Jong-Young Lee, Seung-Whan Lee
ABSTRACT
Sodium bicarbonate has been postulated to prevent contrast-induced acute kidney injury (CI-AKI) by various mechanisms, although the reports are conflicting. We searched MEDLINE, EMBASE, and the Cochrane databases for randomized controlled trials that compared a sodium chloride with a sodium bicarbonate hydration regimen with regard to CI-AKI. Data across 19 clinical trials consisting of 3,609 patients were combined. Preprocedural hydration with sodium bicarbonate was associated with a significant decrease in the rate of CI-AKI (odds ratio [OR] 0.56; 95% confidence interval [CI] 0.36-0.86; P=0.008). Stratified analyses by the type of contrast medium suggested lower odds of CI-AKI with sodium bicarbonate in studies using low-osmolar contrast media (OR 0.40; 95% CI 0.23-0.71, P=0.002) compared with those using the iso-osmolar agents (OR 0.76; 95% CI 0.41-1.43; P=0.40). No significant difference in the rates of postprocedural death (OR 0.49; 95% CI 0.23-1.04; P=0.06) and the requirement for renal replacement therapy (OR 0.94; 95% CI 0.46-1.91; P=0.86) was observed. However, we found significant changes in serum bicarbonate and potassium levels after sodium bicarbonate infusion. This updated meta-analysis demonstrates that sodium bicarbonate-based hydration is superior to sodium chloride in preventing CI-AKI of patients undergoing exposure to iodinated contrast media.
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C