Merck
  • Role of diuretics and lipid formulations in the prevention of amphotericin B-induced nephrotoxicity.
Role of diuretics and lipid formulations in the prevention of amphotericin B-induced nephrotoxicity.
European journal of clinical pharmacology (2013-01-31)
Iman Karimzadeh, Hossein Khalili, Shadi Farsaei, Simin Dashti-Khavidaki, Mohammad Mahdi Sagheb
ABSTRACT
To collect available clinical data to define the role of diuretics and lipid formulations in the prevention of amphotericin B (AmB)-induced nephrotoxicity (AIN) in human populations. A literature search was performed in the following databases: Scopus, Medline, Embase, Cochrane Central Register of Controlled Trials, and Cochrane Database of Systematic Reviews. Co-administration of mannitol failed to show any clinically significant benefit in preventing AIN. Potassium-sparing diuretics, such as amiloride and spironolactone, have been shown to have beneficial effects as an alternative or adjunct to oral/parenteral potassium supplements in preventing hypokalemia due to AmB. Lipid-based formulations of AmB are clinically effective and safe in preventing AIN. However, due to their high cost and limited accessibility, these formulations are generally used as second-line antifungal therapy in cases of conventional AmB refractoriness and/or intolerance or pre-existing renal dysfunction. The potential effects of other nephroprotective agents, such as N-acetylcysteine, AIN merit further considerations and investigations.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Amphotericin B for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard