Merck
  • Molecular and cellular effects of vitamin B12 in brain, myocardium and liver through its role as co-factor of methionine synthase.
Molecular and cellular effects of vitamin B12 in brain, myocardium and liver through its role as co-factor of methionine synthase.
Biochimie (2013-02-19)
Jean-Louis Guéant, Maatem Caillerez-Fofou, Shyuefang Battaglia-Hsu, Jean-Marc Alberto, Jean-Noel Freund, Isabelle Dulluc, Charles Adjalla, Florence Maury, Carole Merle, Jean-Pierre Nicolas, Fares Namour, Jean-Luc Daval
ABSTRACT
Vitamin B12 (cobalamin, cbl) is a cofactor of methionine synthase (MTR) in the synthesis of methionine, the precursor of the universal methyl donor S-Adenosylmethionine (SAM), which is involved in epigenomic regulatory mechanisms. We have established a neuronal cell model with stable expression of a transcobalamin-oleosin chimer and subsequent decreased cellular availability of vitamin B12, which produces reduced proliferation, increased apoptosis and accelerated differentiation through PP2A, NGF and TACE pathways. Anti-transcobalamin antibody or impaired transcobalamin receptor expression produce also impaired proliferation in other cells. Consistently, the transcription, protein expression and activity of MTR are increased in proliferating cells of skin and intestinal epitheliums, in rat intestine crypts and in proliferating CaCo2 cells, while MTR activity correlates with DNA methylation in rat intestine villi. Exposure to nitrous oxide in animal models identified impairment of MTR reaction as the most important metabolic cause of neurological manifestations of B12 deficiency. Early vitamin B12 and folate deprivation during gestation and lactation of a 'dam-progeny' rat model developed in our laboratory is associated with long-lasting disabilities of behavior and memory capacities, with persisting hallmarks related to increased apoptosis, impaired neurogenesis and altered plasticity. We found also an epigenomic deregulation of energy metabolism and fatty acids beta-oxidation in myocardium and liver, through imbalanced methylation/acetylation of PGC-1alpha and decreased expression of SIRT1. These nutrigenomic effects display similarities with the molecular mechanisms of fetal programming. Beside deficiency, B12 loading increases the expression of MTR through internal ribosome entry sites (IRES) and down-regulates MDR-1 gene expression. In conclusion, vitamin B12 influences cell proliferation, differentiation and apoptosis in brain. Vitamin B12 and folate combined deficiency impairs fatty acid oxidation and energy metabolism in liver and heart through epigenomic mechanisms related to imbalanced acetylation/methylation. Some but not all of these effects reflect the upstream role of vitamin B12 in SAM synthesis.
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cyanocobalamin (B12), analytical standard
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard