Merck
  • Nickel and cobalt-catalyzed coupling of alkyl halides with alkenes via heck reactions and radical conjugate addition.
Nickel and cobalt-catalyzed coupling of alkyl halides with alkenes via heck reactions and radical conjugate addition.
Mini reviews in medicinal chemistry (2013-04-03)
Qun Qian, Zhenhua Zang, Yang Chen, Weiqi Tong, Hegui Gong
ABSTRACT
Cross-coupling of alkyl halides with alkenes leading to Heck-type and addition products is summarized. The development of Heck reaction with aliphatic halides although has made significant progress in the past decade and particularly recently, it was much less explored in comparison with the aryl halides. The use of Ni- and Co-catalyzed protocols allowed efficient Heck coupling of activated and unactivated alkenes with 1°, 2° and 3° alkyl halides. In addition, radical conjugate addition to activated alkenes has become a well-established method that has led to efficient construction of many natural products. The utilization of Ni- and Co-catalyzed strategies would avoid toxic tin reagents, and therefore worth exploring. The recent development of Ni- and Co-catalyzed addition of alkyl halides to alkenes displays much improved reactivity and functional group tolerance. In this mini-review, we also attempt to overview the mechanisms that are proposed in the reactions, aiming at providing insight into the nickel and cobalt-catalyzed coupling of alkyl halides with alkenes.
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Nickel, pellets, 50g, max. size 10mm, 99.999%
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 50x50mm, thickness 0.025mm, 99.9%
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Cobalt, wire reel, 2m, diameter 0.05mm, hard, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Nickel, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Nickel, foil, light tested, 100x100mm, thickness 0.025mm, 99.9%
Nickel, wire reel, 5m, diameter 0.025mm, as drawn, 99.98%
Nickel, foil, 5m coil, thickness 0.05mm, annealed, 99%
Nickel, wire reel, 0.1m, diameter 0.75mm, as drawn, 99.99+%
Nickel, foil, 0.5m coil, thickness 0.025mm, 99.9%
Nickel, rod, 500mm, diameter 2.0mm, 99.99+%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Nickel, foil, 0.5m coil, thickness 0.0075mm, 99.95%