Merck
Toxicology of cadmium and its damage to mammalian organs.
Metal ions in life sciences (2013-02-23)
Frank Thévenod, Wing-Kee Lee
ABSTRACT
The detrimental health effects of cadmium (Cd) were first described in the mid 19th century. As part of industrial developments, increasing usage of Cd has led to widespread contamination of the environment that threatens human health, particularly today. Rather than acute, lethal exposures, the real challenge in the 21st century in a global setting seems to be chronic low Cd exposure (CLCE), mainly from dietary sources. Ubiquity of Cd makes it a serious environmental health problem that needs to be thoroughly assessed because it already affects or will affect large proportions of the world's population. CLCE is a health problem that affects increasingly organ toxicity, especially nephrotoxicity, without a known threshold, implying that there is currently no safe limit for CLCE. In this chapter, we summarize current knowledge on the sources of Cd in the environment, describe the entry pathways for Cd into mammalian organisms, sum up the major organs targeted by acute or chronic Cd exposure and review the impact of Cd on organ function and human health. We also aim to put early pioneering studies on Cd poisoning into perspective in the context of recent ground-breaking prospective long-term population studies, which link CLCE to leading causes of diseases in modern societies - cancer, diabetes, and cardiovascular diseases, and of state-of-the-art studies detailing cellular and molecular mechanisms of acute and chronic Cd toxicity.
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 50mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Cadmium, foil, 50mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, wire reel, 0.1m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 50mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, rod, 200mm, diameter 5.0mm, as drawn, 99.99+%
Cadmium, foil, light tested, 100x100mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 6mm disks, thickness 0.005mm, 99.7+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, foil, light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 6mm disks, thickness 0.015mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%