Merck
Dosing and antipyretic efficacy of oral acetaminophen in children.
Clinical therapeutics (2013-08-27)
Anthony R Temple, Brigham R Temple, Edwin K Kuffner
ABSTRACT
A standardized approach to dosing acetaminophen in pediatric populations was published in 1983. That review proposed specific weight-related dosing for infants and children weighing 6 through 95 lb and an age-based schedule for children aged <4 months through 11 years. Subsequent clinical studies evaluating these and alternative doses of acetaminophen supported the recommended 10-15-mg/kg dose. This article reviewed published and unpublished pediatric antipyretic data to provide a critical assessment of the 10-15-mg/kg oral dose and the current pediatric oral dosing schedules for acetaminophen. Published literature and unpublished clinical trials that evaluated the antipyretic efficacy of acetaminophen in children were reviewed. The PubMed database was searched using the term acetaminophen or paracetamol, with study criteria limited to randomized, controlled trials; oral dosing; patient age <12 years; and publication between 1982 and August 2012. All of the sponsor's unpublished antipyretic clinical studies completed between 1980 and August 2012 and involving at least 1 oral-formulation acetaminophen-only treatment arm were identified. Data from published literature containing sufficient detail to verify doses; dosing frequency; and, when necessary, estimates from figures, and from acetaminophen arms of the unpublished studies were analyzed. Thirteen unpublished trials enrolled 705 children to receive an oral dose of 10-15 mg/kg of acetaminophen. This dose resulted in a rapid onset of temperature reduction, with a maximum temperature decrement of ~3 hours following administration. Results from 40 published clinical trials in which 2332 children received oral acetaminophen for fever support these findings. The most common adverse events reported in any of the reported studies were gastrointestinal in nature and generally mild in intensity. Data support the recommended 10-15-mg/kg oral dose and demonstrate that the age and weight schedules for over-the-counter acetaminophen proposed in 1983 remain appropriate.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Paracetamol, European Pharmacopoeia (EP) Reference Standard